Silica gel 12-24 mesh for drying solvents
Supplier: STREM CHEMICALS INC MS
93-1424-2000G
93-1424-500G
100199-492EA
227.88
USD
100199-492
100210-192
Silica gel 12-24 mesh for drying solvents
Silica gel
Formula:
SiO₂ MW: 60.08 g/mol Boiling Pt: 2230 Melting Pt: 1710 |
MDL Number:
MFCD00011232 CAS Number: 63231-67-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...