Silica gel 6-16 mesh, with indicator

Supplier: STREM CHEMICALS INC MS

93-1423-500G 93-1423-2000G
100198-310EA 165.6 USD
100198-310 100201-064
Silica gel 6-16 mesh, with indicator
Silica gel
Formula: SiO₂
MW: 60.08 g/mol
MDL Number: MFCD00011232
CAS Number: 63231-67-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR