Aluminium triethanolate ≥99%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Aluminumethoxide, Aluminum ethoxide, Aluminium triethylate

93-1370-100G 93-1370-25G
100201-898EA 300.06 USD
100201-898 100201-900
Aluminium triethanolate ≥99%
Aluminium triethanolate

Alkoxide, Metal Alkoxides, Volatile Precursors for CVD

Formula: C₆H₁₅AlO₃
MW: 162.16
Boiling Pt: 210 °C (10 mmHg)
Melting Pt: 130 °C
MDL Number: MFCD00009658
CAS Number: 555-75-9
UN: 2925
ADR: 4.1 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR