Aluminium stearate
Supplier: STREM CHEMICALS INC MS
93-1349-1000G
93-1349-5000G
100205-314EA
77.85
USD
100205-314
100203-380
Aluminium stearate
Aluminium stearate
Formula:
C₅₄H₁₀₅AlO₆ Melting Pt: 103 °C Density: 1,010 |
MDL Number:
MFCD00036389 CAS Number: 637-12-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...