Bis(2-ethylhexanoato)hydroxyaluminum ≥96%

Supplier: STREM CHEMICALS INC MS
Warning

93-1326-500G 93-1326-100G
100204-170EA 448.68 USD
100204-170 100210-180
Bis(2-ethylhexanoato)hydroxyaluminum ≥96%
Bis(2-ethylhexanoato)hydroxyaluminum
Formula: C₁₆H₃₁AlO₅
Flash Pt: >350 °F
MDL Number: MFCD00041940
CAS Number: 30745-55-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR