Isopropylmagnesium chloride 2-3M in ether

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: i-Propylmagnesium chloride

93-1271-250MMOL 93-1271-1MOL
100207-772EA 134.46 USD
100207-772 100209-008
Isopropylmagnesium chloride 2-3M in ether
Isopropylmagnesium chloride

Grignard

Formula: C₃H₇ClMg
MW: 102.84 g/mol
MDL Number: MFCD00000466
CAS Number: 1068-55-9
UN: 3399
ADR: 4.3,I

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR