Bis(trifluoro-2,4-pentanedionato)magnesium(II) hydrate ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Magnesium trifluoroacetylacetonate dihydrate

93-1259-25G 93-1259-5G
100207-456EA 644 USD
100207-456 100199-478
Bis(trifluoro-2,4-pentanedionato)magnesium(II) hydrate ≥98%
Bis(trifluoro-2,4-pentanedionato)magnesium(II) hydrate
Formula: C₁₀H₈F₆MgO₄
MW: 330.47 g/mol
MDL Number: MFCD00077561
CAS Number: 53633-79-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR