Magnesium naphthenate (3,5 - 8,0% Mg)

Supplier: STREM CHEMICALS INC MS

93-1236-500G 93-1236-100G
100201-880EA 546.34 USD
100201-880 100204-158
Magnesium naphthenate (3,5 - 8,0% Mg)
Magnesium naphthenate
MDL Number: MFCD00147520
CAS Number: 68424-71-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR