Magnesium molybdenum oxide ≥99.9% (Mo basis)

Supplier: STREM CHEMICALS INC MS

Synonyms: Magnesium molybdate

93-1235-50G 93-1235-10G
100206-922EA 174.09 USD
100206-922 100201-878
Magnesium molybdenum oxide ≥99.9% (Mo basis)
Magnesium molybdenum oxide
Formula: MgMoO₄
MW: 184.243 g/mol
Density: 2.208
MDL Number: MFCD00016195
CAS Number: 13767-03-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR