Magnesium carbonate hydroxide pentahydrate ≥99%

Supplier: STREM CHEMICALS INC MS

Synonyms: Magnesium carbonate basic pentahydrate, Magnesium hydroxide carbonate pentahydrate

93-1220-500G 93-1220-100G
100204-154EA 104.74 USD
100204-154 100201-872
Magnesium carbonate hydroxide pentahydrate ≥99%
Magnesium carbonate hydroxide pentahydrate
Formula: C₄H₂Mg₅O₁₄·5H₂O
MW: 485.65 g/mol (395.58 anhy)
MDL Number: MFCD00149784
CAS Number: 56378-72-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR