Ethylmagnesium bromide MMMA Stabilised with diethyl ether

Supplier: STREM CHEMICALS INC MS
Danger

93-1212-250MMOL 93-1212-1MOL
100205-296EA 77.85 USD
100205-296 100208-496
Ethylmagnesium bromide MMMA Stabilised with diethyl ether
Ethylmagnesium bromide

Grignard

Formula: C₂H₅BrMg
MDL Number: MFCD00000043
CAS Number: 925-90-6
UN: 3399
ADR: 4.3,I

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR