Butylmagnesium chloride 1.5-3.0 M in THF

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: n-Butylmagnesium chloride

93-1204-250MMOL 93-1204-1MOL
100196-474EA 124.55 USD
100196-474 100206-916
Butylmagnesium chloride 1.5-3.0 M in THF
Butylmagnesium chloride

Grignard

Formula: C₄H₉ClMg
MW: 116.87 g/mol
Flash Pt: 1 °F
MDL Number: MFCD00000475
CAS Number: 693-04-9
UN: 3399
ADR: 4.3,I

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR