Sodium hexacyanoferrate(II) decahydrate

Supplier: STREM CHEMICALS INC MS

Synonyms: Sodium ferrocyanide decahydrate

93-1140-500G 93-1140-2000G
100204-144EA 232.12 USD
100204-144 100203-360
Sodium hexacyanoferrate(II) decahydrate
Sodium hexacyanoferrate(II) decahydrate
Formula: C₆H₂₀FeN₆Na₄O₁₀
MW: 484.062 g/mol
Density: 1,458
MDL Number: MFCD00085360
CAS Number: 14434-22-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR