Sodium 2,4-pentanedionate hydrate ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

93-1106-10G 93-1106-50G
101419-156EA 144.37 USD
101419-156 100198-954
Sodium 2,4-pentanedionate hydrate ≥98%
Sodium 2,4-pentanedionate hydrate
Formula: C₅H₇NaO₂·xH₂O
Melting Pt: 210 °C (dec.)
MDL Number: MFCD00078034
CAS Number: 86891-03-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR