Sodium bis(2-methoxyethoxy)aluminium hydride ≥70% in toluene

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Sodium dihydrobis(2-methoxyethoxy)aluminate, SBAH, Sodium bis(2-methoxyethoxy)aluminum dihydride

93-1077-500G 93-1077-100G
100197-664EA 159.94 USD
100197-664 100201-852
Sodium bis(2-methoxyethoxy)aluminium hydride ≥70% in toluene
Sodium bis(2-methoxyethoxy)aluminium hydride
Formula: C₆H₁₆AlNaO₄
MW: 202.16 g/mol
Density: 1.034 (20°)
Flash Pt: 40 °F
MDL Number: MFCD00011631
CAS Number: 22722-98-1
UN: 3148
ADR: 4.3 I

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR