2,4,6-Trimethoxyboroxin ≥95%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Trimethoxycyclotriboroxane, Trimethoxyboroxine

93-0574-100G 93-0574-500G
100196-440EA 62.28 USD
100196-440 100199-474
2,4,6-Trimethoxyboroxin ≥95%
2,4,6-Trimethoxyboroxin
Formula: C₃H₉B₃O₆
MW: 173.53 g/mol
Melting Pt: 10…11 °C
Density: 1,216
Flash Pt: 50 °F
MDL Number: MFCD00005946
CAS Number: 102-24-9
UN: 1993
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR