Lithium tetraphenylborate tris(1,2-dimethoxyethane) adduct ≥98%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
93-0572-1G
93-0572-5G
100198-946EA
72.18
USD
100198-946
100205-262
Lithium tetraphenylborate tris(1,2-dimethoxyethane) adduct ≥98%
Lithium tetraphenylborate tris(1,2-dimethoxyethane) adduct
Formula:
C₂₄H₂₀BLi.3C₄H₁₀O₂ MW: 596.531 g/mol |
MDL Number:
MFCD00013311 CAS Number: 75965-35-4 UN: 1325 ADR: 4.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...