Triphenylborane ≥95%

Supplier: STREM CHEMICALS INC MS
Danger

93-0543-1G 93-0543-25G 93-0543-5G
100204-128EA 155.69 USD
100204-128 100206-896 100206-898
Triphenylborane ≥95%
Triphenylborane
Formula: C₁₈H₁₅B
MW: 242.13 g/mol
Boiling Pt: 208 °C (14 mmHg)
Melting Pt: 146…148 °C
MDL Number: MFCD00003007
CAS Number: 960-71-4
UN: 1325
ADR: 4.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR