Triethanolamine borate ≥95%

Supplier: STREM CHEMICALS INC MS

Synonyms: 2,2',2''-Nitrilotriethyl Borate

93-0538-100G 93-0538-25G
100208-076EA 144.37 USD
100208-076 100201-822
Triethanolamine borate ≥95%
Triethanolamine borate
Formula: C₆H₁₂BNO₃
MW: 156.98 g/mol
Melting Pt: 235…237 °C
MDL Number: MFCD00003272
CAS Number: 283-56-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR