Dichlorophenylborane ≥97%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Phenylboron dichloride

93-0533-25G 93-0533-5G 93-0533-1G
100209-276EA 488.31 USD
100209-276 100201-048 100206-894
Dichlorophenylborane ≥97%
Dichlorophenylborane
Formula: C₆H₅BCl₂
MW: 158.82 g/mol
Boiling Pt: 175 °C
Melting Pt: 7 °C
Density: 1.194 (20°)
Flash Pt: –5 °F
MDL Number: MFCD00013612
CAS Number: 873-51-8
UN: 2924
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR