Lithium bromide hydrate ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Lithium bromide xhydrate

93-0371-250G 93-0371-50G
100204-124EA 108.98 USD
100204-124 100210-124
Lithium bromide hydrate ≥99%
Lithium bromide hydrate
Formula: BrLi·xH₂O
MW: 86.85 g/mol (anhy)
Boiling Pt: 1265
Melting Pt: 549
MDL Number: MFCD00149763
CAS Number: 85017-82-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR