Methyllithium (1.5M) in ethyl ether with LiBr
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Methyllithium - lithium bromide complex, Methyllithium solution
93-0358-1MOL
93-0358-250MMOL
100196-426EA
653.91
USD
100196-426
100199-470
Methyllithium (1.5M) in ethyl ether with LiBr
Methyllithium
Formula:
CH₃Li MW: 21.975 g/mol |
MDL Number:
MFCD00008253 CAS Number: 917-54-4 UN: 3394 ADR: 4.2,I |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...