Lithium dimethylamide ≥95%

Supplier: STREM CHEMICALS INC MS
Danger

93-0318-5G 93-0318-25G
100206-874EA 169.85 USD
100206-874 100210-672
Lithium dimethylamide ≥95%
Lithium dimethylamide
Formula: C₂H₆LiN
Melting Pt: Dec.
MDL Number: MFCD00008295
CAS Number: 3585-33-9
UN: 2846
ADR: 4.2,I

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR