Bismuth (III) trifluoromethanesulfonate ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Bismuth triflate, Bismuth(III) trifluoromethanesulphonate

83-8000-1G 83-8000-25G 83-8000-5G
100201-782EA 45.29 USD
100201-782 100201-784 100201-786
Bismuth (III) trifluoromethanesulfonate ≥98%
Bismuth(III) trifluoromethanesulphonate

Metal Triflate

Formula: C₃BiF₉O₉S₃
MW: 656.19 g/mol
MDL Number: MFCD02093669
CAS Number: 88189-03-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR