Bismuth(III) 2-ethylhexanoate ≥72% in mineral spirits (~28% Bi)
Supplier: STREM CHEMICALS INC MS
Synonyms:
Bismuth 2-ethylhexanoate
83-2400-1000G
83-2400-250G
100205-852EA
242.03
USD
100205-852
100210-108
Bismuth(III) 2-ethylhexanoate ≥72% in mineral spirits (~28% Bi)
Bismuth (III) 2-ethylhexanoate
Formula:
C₂₄H₄₅BiO₆ MW: 638.598 g/mol Density: 1.28 Flash Pt: 162 °F |
MDL Number:
MFCD00043803 CAS Number: 67874-71-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...