Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')bismuth(III) ≥98% (99.9%-Bi)

Supplier: STREM CHEMICALS INC MS

83-1000-5G 83-1000-1G 83-1000-25G
100211-190EA 317.05 USD
100211-190 100211-186 100211-188
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')bismuth(III) ≥98% (99.9%-Bi)
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')bismuth(III)

Metal Beta-diketonates, Metal TMHD, Volatile Precursors for CVD

Formula: C₃₃H₅₇BiO₆
Boiling Pt: 295 °C/ 150 °C (0.05 mmHg) (dec./ subl.)
Melting Pt: 114…116 °C
MDL Number: MFCD00064763
CAS Number: 142617-53-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR