Bis(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')lead(II) ≥99%

Supplier: STREM CHEMICALS INC MS

Synonyms: Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)lead(II), Pb(TMHD)2

82-2100-25G 82-2100-5G 82-2100-1G
100196-408EA 837.91 USD
100196-408 100196-410 100201-776
Bis(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')lead(II) ≥99%
Bis(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')lead(II)

Metal Beta-diketonates, Metal TMHD, Volatile Precursors for CVD

Formula: C₂₂H₃₈O₄Pb
MW: 573.73 g/mol
Boiling Pt: 325 °C/ 134 °C (0.1 mmHg) (dec./ subl.)
Melting Pt: 126…128 °C
MDL Number: MFCD00070468
CAS Number: 21319-43-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR