Thallium hydrotris[3-(2-pyridyl)pyrazol-1-yl]borate ≥98%

Supplier: STREM CHEMICALS INC MS
Danger

81-5000-250MG 81-5000-1G
100201-042EA 67.94 USD
100201-042 100196-974
Thallium hydrotris[3-(2-pyridyl)pyrazol-1-yl]borate ≥98%
Thallium hydrotris[3-(2-pyridyl)pyrazol-1-yl]borate
Melting Pt: 283…285 °C
MDL Number: MFCD02684575
CAS Number: 165257-90-9
UN: 1707
ADR: 6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR