Thallium hydrotris(indazol-2-yl)borate ≥98%

Supplier: STREM CHEMICALS INC MS
Danger

81-4500-250MG 81-4500-1G
100203-330EA 152.86 USD
100203-330 100209-486
Thallium hydrotris(indazol-2-yl)borate ≥98%
Thallium hydrotris(indazol-2-yl)borate
Melting Pt: 254…256 °C
MDL Number: MFCD02684574
CAS Number: 162978-03-2
UN: 1707
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR