(2,2,6,6-Tetramethyl-3,5-heptanedionato-O,O')thallium(I) ≥99%

Supplier: STREM CHEMICALS INC MS
Danger

81-1000-5G 81-1000-1G
100209-394EA 574.65 USD
100209-394 100209-392
(2,2,6,6-Tetramethyl-3,5-heptanedionato-O,O')thallium(I) ≥99%
(2,2,6,6-Tetramethyl-3,5-heptanedionato-O,O')thallium(I)

Metal Beta-diketonates, Metal TMHD, Volatile Precursors for CVD

Formula: C₁₁H₁₉TlO₂
Boiling Pt: 260 °C/ 110 °C (0.1 mmHg) (dec./ subl.)
Melting Pt: 159…164 °C
MDL Number: MFCD00070465
CAS Number: 56713-38-3
UN: 1707
ADR: 6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR