Mercury(II) trifluoromethanesulfonate ≥98%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Mercury(II) trifluoromethanesulphonate, Mercury triflate
80-2200-25G
80-2200-5G
100201-254EA
625.6
USD
100201-254
100208-062
Mercury(II) trifluoromethanesulfonate ≥98%
Mercury(II) trifluoromethanesulphonate
Metal Triflate
Formula:
C₂F₆HgO₆S₂ MW: 498.73 g/mol |
MDL Number:
MFCD00144746 CAS Number: 49540-00-3 UN: 2025 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...