Methyl(triphenylphosphine)gold(I) ≥99%

Supplier: STREM CHEMICALS INC MS

79-5000-250MG 79-5000-1G
101324-832EA 196.74 USD
101324-832 101324-834
Methyl(triphenylphosphine)gold(I) ≥99%
Methyl(triphenylphosphine)gold(I)
Formula: C₁₉H₁₉AuP
MW: 475.2945 g/mol
Melting Pt: ~150 °C (dec.)
MDL Number: MFCD07369042
CAS Number: 23108-72-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR