Tris[triphenylphosphinegold(I)]oxonium tetrafluoroborate ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

79-3600-250MG 79-3600-1G
100201-036EA 142.95 USD
100201-036 100196-970
Tris[triphenylphosphinegold(I)]oxonium tetrafluoroborate ≥98%
Tris[triphenylphosphinegold(I)]oxonium tetrafluoroborate
Formula: C₅₄H₄₈Au₃B₄F₁₆OP₃
MW: 1743.9977 g/mol
Melting Pt: 207 °C (dec.)
MDL Number: MFCD06658141
CAS Number: 53317-87-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR