Potassium tetrabromoaurate(III) dihydrate ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

79-3250-1G 79-3250-5G
100199-458EA 167.02 USD
100199-458 100207-448
Potassium tetrabromoaurate(III) dihydrate ≥99%
Potassium tetrabromoaurate(III) dihydrate
Formula: KAuBr₄.2H₂O
MDL Number: MFCD00049650
CAS Number: 14323-32-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR