Hydrogen tetrabromoaurate(III) hydrate ≥99.9% (Au basis)
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Bromoauric acid
79-2150-1G
79-2150-5G
100200-002EA
167.02
USD
100200-002
100208-968
Hydrogen tetrabromoaurate(III) hydrate ≥99.9% (Au basis)
Hydrogen tetrabromoaurate(III) hydrate
Formula:
HAuBr₄·xH₂O MW: 517.61 g/mol (anhy) |
MDL Number:
MFCD00150570 CAS Number: 17083-68-0 UN: 1759 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...