Dimethyl(trifluoroacetylacetonate)gold(III) ≥98% (99.9%-Au)

Supplier: STREM CHEMICALS INC MS

79-1600-500MG 79-1600-2G
100203-322EA 2675.08 USD
100203-322 100203-324
Dimethyl(trifluoroacetylacetonate)gold(III) ≥98% (99.9%-Au)
Dimethyl(trifluoroacetylacetonate)gold(III)

Metal Beta-diketonates, Volatile Precursors for CVD

MDL Number: MFCD01631288
CAS Number: 63470-53-1
UN: 2811
ADR: 6.1 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR