(Triphenylphosphine)gold(I) chloride ≥98% (99.9+%-Au)

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Chlorotriphenylphosphine gold(I), Chloro(triphenylphosphine)gold(I), Triphenylphosphinechlorogold

79-1000-1G 79-1000-5G
100206-840EA 215.14 USD
100206-840 100206-842
(Triphenylphosphine)gold(I) chloride ≥98% (99.9+%-Au)
(Triphenylphosphine)gold(I) chloride
Formula: C₁₈H₁₅AuClP
MDL Number: MFCD00009588
CAS Number: 14243-64-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR