Chlorotri-t-butylphosphinegold(I) ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

79-0740-250MG 79-0740-1G
101419-144EA 175.51 USD
101419-144 101419-146
Chlorotri-t-butylphosphinegold(I) ≥99%
Chlorotri-t-butylphosphinegold(I)
Formula: C₁₂H₂₈AuClP
MW: 435.7440 g/mol
MDL Number: MFCD09753040
CAS Number: 69550-28-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR