Gold, nanoparticles (water soluble) [AuNP: 11-20 nm (Gelatin)]

Supplier: STREM CHEMICALS INC MS
Warning

79-0126-100ML 79-0126-25ML
101442-272EA 1082.77 USD
101442-272 101442-274
Gold, nanoparticles (water soluble) [AuNP: 11-20 nm (Gelatin)]
Gold

Nanomaterials

Formula: Au
MW: 196.97 g/mol
MDL Number: MFCD00003436
CAS Number: 7440-57-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR