[mu-Bis(diphenylphosphino)methane]dichlorodigold(I) ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

79-0115-250MG 79-0115-1G
101442-200EA 205.23 USD
101442-200 101442-202
[mu-Bis(diphenylphosphino)methane]dichlorodigold(I) ≥99%
[mu-Bis(diphenylphosphino)methane]dichlorodigold(I)
MDL Number: MFCD08705280
CAS Number: 37095-27-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR