Gold, nanoparticles (water soluble) [AuNP: 10 nm (Citrate)]

Supplier: STREM CHEMICALS INC MS
Warning

79-0114-25ML 79-0114-100ML
101442-270EA 274.58 USD
101442-270 101442-268
Gold, nanoparticles (water soluble) [AuNP: 10 nm (Citrate)]
Gold

Nanomaterials

Formula: Au
MW: 196.97 g/mol
MDL Number: MFCD00003436
CAS Number: 7440-57-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR