Gold, nanoparticles (water soluble) [AuNP: 12-16 nm (Starch)]
Supplier: STREM CHEMICALS INC MS
|
Warning
|
79-0108-100ML
79-0108-25ML
101442-294EA
1023.32
USD
101442-294
101442-296
Gold, nanoparticles (water soluble) [AuNP: 12-16 nm (Starch)]
Gold
Nanomaterials
Formula:
Au MW: 196.97 g/mol |
MDL Number:
MFCD00003436 CAS Number: 7440-57-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...