Tetraammineplatinum(II) tetrachloroplatinate(II) ≥99%

Supplier: STREM CHEMICALS INC MS

Synonyms: Tetraammineplatinum tetrachloroplatinate(II)

78-2015-1G 78-2015-5G
100210-670EA 338.28 USD
100210-670 100199-998
Tetraammineplatinum(II) tetrachloroplatinate(II) ≥99%
Tetraammineplatinum(II) tetrachloroplatinate(II)
Formula: [Pt(NH₃)₄][PtCl₄]
MW: 600.102 g/mol
MDL Number: MFCD01459899
CAS Number: 13820-46-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR