Tetraammineplatinum(II) nitrate ≥99%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
78-2010-5G
78-2010-250MG
78-2010-1G
100200-496EA
891.69
USD
100200-496
101442-472
100200-494
Tetraammineplatinum(II) nitrate ≥99%
Tetraammineplatinum(II) nitrate
Formula:
H₁₂N₆O₆Pt MW: 387.216 g/mol |
MDL Number:
MFCD00011624 CAS Number: 20634-12-2 UN: 1477 ADR: 5.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...