Tetraammineplatinum(II) hydroxide hydrate (59% Pt)

Supplier: STREM CHEMICALS INC MS

78-2005-1G 78-2005-5G
100208-688EA 334.03 USD
100208-688 100208-690
Tetraammineplatinum(II) hydroxide hydrate (59% Pt)
Tetraammineplatinum(II) hydroxide hydrate
Formula: H₁₄N₄O₂Pt·xH₂O
MW: 297.23 g/mol (anhy)
MDL Number: MFCD00151253
CAS Number: 312695-70-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR