Tetraammineplatinum(II) chloride hydrate ≥99% (99.95%-Pt)
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
AKOS015924294, AC-24209
78-2000-1G
78-2000-5G
100198-898EA
234.95
USD
100198-898
100206-130
Tetraammineplatinum(II) chloride hydrate ≥99% (99.95%-Pt)
Tetraammineplatinum(II) chloride hydrate
Formula:
Pt(NH₃)₄Cl₂·xH₂O MW: 334.11 g/mol (anhy) |
MDL Number:
MFCD00149947 CAS Number: 108374-32-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...