Potassium trichloro(ethylene)platinate(II) monohydrate

Supplier: STREM CHEMICALS INC MS
Warning

78-1990-2G 78-1990-500MG
100199-454EA 621.35 USD
100199-454 100198-262
Potassium trichloro(ethylene)platinate(II) monohydrate
Potassium trichloro(ethylene)platinate(II) monohydrate
Formula: C₂H₄Cl₃KPt·H₂O
MW: 386.61 g/mol (368.59 anhy)
MDL Number: MFCD00150524
CAS Number: 16405-35-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR