Platinum sulfite acid solution (15.3% Pt)

Supplier: STREM CHEMICALS INC MS
Danger

78-1925-5G 78-1925-25G
100208-686EA 583.14 USD
100208-686 100208-684
Platinum sulfite acid solution (15.3% Pt)
Platinum sulfite acid
MDL Number: MFCD00058877
CAS Number: 61420-92-6
UN: 1760
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR