Platinum 5% on alumina
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Platinium
78-1660-50G
78-1660-10G
100197-616EA
897.35
USD
100197-616
100204-110
Platinum 5% on alumina
Platinum
Formula:
Pt MW: 195.08 g/mol Boiling Pt: 3827 Melting Pt: 1769 Density: 21.45 |
MDL Number:
MFCD00011179 CAS Number: 7440-06-4 Merck Index: 12,07689 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...