Platinum 0,5%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Platinium

78-1640-100G 78-1640-25G
100196-966EA 421.78 USD
100196-966 100211-174
Platinum 0,5%
Platinum
Formula: Pt
MW: 195.08 g/mol
Boiling Pt: 3827
Melting Pt: 1769
Density: 21.45
MDL Number: MFCD00011179
CAS Number: 7440-06-4
Merck Index: 12,07689

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR